Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012263.4 | NP_036395.1 | 9 | 1161 | ||
NR_027779.1 | 10 | 1265 | ||||
XM_006724225.3 | XP_006724288.1 | 9 | 1127 | |||
XM_011530108.2 | XP_011528410.1 | 10 | 1188 | |||
XM_011530110.2 | XP_011528412.1 | 9 | 1148 | |||
XM_017028751.1 | XP_016884240.1 | 10 | 1163 | |||
XM_017028752.1 | XP_016884241.1 | 9 | 1074 | |||
XM_017028753.1 | XP_016884242.1 | 10 | 1263 | |||
GenBank mRNA | AF104927.1 | - | 9 | 1062 | ||
AF173935.1 | - | 10 | 1175 | |||
AK095996.1 | - | 9 | 1137 | |||
AK315658.1 | - | 9 | 1086 | |||
AL096883.1 | - | 9 | 1081 | |||
AL096886.1 | - | 8 | 1009 | |||
AL136687.1 | - | 9 | 1161 | |||
BC014968.1 | - | 9 | 1148 | 4876418 | ||
BG911167.1 | - | 3 | 189 | |||
CR456599.1 | - | 7 | 924 |