Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 8 | 931 | ||
GenBank mRNA | AF151809.1 | - | 8 | 843 | ||
AK001087.1 | - | 8 | 870 | |||
AK097015.1 | - | 1 | 1546 | |||
AK222657.1 | - | 8 | 805 | |||
AK312773.1 | - | 8 | 766 | |||
AY189688.1 | - | 8 | 846 | |||
BC007830.1 | - | 8 | 838 | 3635175 | ||
BC011681.2 | - | 8 | 891 | 3010765 | ||
BC015200.1 | - | 8 | 781 | 4333051 | ||
CR456483.1 | - | 8 | 743 |