Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082967.2 | NP_001076436.1 | 2 | 272 | ||
NM_015381.6 | NP_056196.2 | 2 | 293 | |||
GenBank mRNA | AF131851.1 | - | 2 | 238 | ||
AK123443.1 | - | 3 | 232 | |||
AK125256.1 | - | 2 | 293 | |||
AY325118.1 | - | 2 | 267 | |||
AY358847.1 | - | 3 | 168 | |||
BC039396.1 | - | 2 | 251 | 5299791 | ||
DB481230.1 | - | 2 | 272 |