Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135004.2 | NP_001128476.2 | 3 | 530 | ||
NM_001135005.2 | NP_001128477.1 | 2 | 737 | |||
NM_012266.5 | NP_036398.3 | 2 | 385 | |||
XM_005251428.3 | XP_005251485.1 | 2 | 304 | |||
XM_006716751.2 | XP_006716814.1 | 3 | 394 | |||
XM_011517842.2 | XP_011516144.1 | 3 | 971 | |||
GenBank mRNA | AF087870.1 | - | 3 | 387 | ||
AF088982.1 | - | 2 | 288 | |||
AK023253.1 | - | 2 | 281 | |||
AK299647.1 | - | 2 | 737 | |||
AK314671.1 | - | 2 | 262 | |||
AL536638.3 | - | 3 | 418 | |||
BI915181.1 | - | 2 | 689 | |||
BU845478.1 | - | 2 | 298 | |||
BX419166.2 | - | 2 | 726 |