Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135004.2 | NP_001128476.2 | 4 | 1103 | ||
NM_001135005.2 | NP_001128477.1 | 3 | 1310 | |||
NM_012266.5 | NP_036398.3 | 3 | 958 | |||
XM_005251428.3 | XP_005251485.1 | 3 | 877 | |||
XM_006716751.2 | XP_006716814.1 | 4 | 967 | |||
XM_011517842.2 | XP_011516144.1 | 4 | 1544 | |||
GenBank mRNA | AF087870.1 | - | 4 | 960 | ||
AF088982.1 | - | 3 | 861 | |||
AK023253.1 | - | 3 | 854 | |||
AK299647.1 | - | 3 | 1310 | |||
AK314671.1 | - | 3 | 835 | |||
BU845478.1 | - | 4 | 758 |