Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135004.2 | NP_001128476.2 | 5 | 1229 | ||
NM_001135005.2 | NP_001128477.1 | 4 | 1436 | |||
NM_012266.5 | NP_036398.3 | 4 | 1084 | |||
XM_005251428.3 | XP_005251485.1 | 4 | 1003 | |||
XM_006716751.2 | XP_006716814.1 | 5 | 1093 | |||
XM_011517842.2 | XP_011516144.1 | 5 | 1670 | |||
GenBank mRNA | AF087870.1 | - | 5 | 1086 | ||
AF088982.1 | - | 4 | 987 | |||
AK023253.1 | - | 4 | 980 | |||
AK299647.1 | - | 4 | 1436 | |||
AK314671.1 | - | 4 | 961 |