Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171713.1 | NP_001165184.1 | 6 | 406 | ||
NM_012157.3 | NP_036289.3 | 6 | 409 | |||
XM_005265014.2 | XP_005265071.1 | 6 | 429 | |||
XM_005265015.3 | XP_005265072.1 | 6 | 630 | |||
XM_005265016.2 | XP_005265073.1 | 6 | 374 | |||
XM_005265017.3 | XP_005265074.1 | 6 | 406 | |||
XM_011533559.2 | XP_011531861.1 | 7 | 575 | |||
XM_017006092.1 | XP_016861581.1 | 7 | 467 | |||
XM_017006093.1 | XP_016861582.1 | 6 | 457 | |||
XM_017006094.1 | XP_016861583.1 | 7 | 490 | |||
GenBank mRNA | AF174589.1 | - | 7 | 380 | ||
AF176518.1 | - | 6 | 344 | |||
AF186273.1 | - | 6 | 359 | |||
AK001271.1 | - | 6 | 409 | |||
AK001438.1 | - | 6 | 377 | |||
AK294209.1 | - | 3 | 160 | |||
AK297534.1 | - | 6 | 374 | |||
AK298967.1 | - | 6 | 375 | |||
AK299665.1 | - | 7 | 492 | |||
AK302218.1 | - | 8 | 637 | |||
AK303764.1 | - | 4 | 269 | |||
AK316229.1 | - | 6 | 430 | |||
AK316383.1 | - | 7 | 549 | |||
AU124816.1 | - | 6 | 409 | |||
BC031556.1 | - | 6 | 353 | 5197151 | ||
CR457121.1 | - | 6 | 338 |