Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014351.3 | NP_055166.1 | 5 | 690 | ||
XM_011530121.1 | XP_011528423.1 | 2 | 295 | |||
GenBank mRNA | AF115311.1 | - | 5 | 574 | ||
AF176342.1 | - | 5 | 594 | |||
AF188698.1 | - | 5 | 657 | |||
AF251263.1 | - | 5 | 603 | |||
AK294622.1 | - | 2 | 287 | |||
AK295399.1 | - | 5 | 523 | |||
AK313048.1 | - | 5 | 615 | |||
AL590119.1 | - | 5 | 574 | |||
BC022459.1 | - | 5 | 583 | 4795827 | ||
BC028171.1 | - | 5 | 662 | 5243984 | ||
BC030665.1 | - | 5 | 718 | 5258865 | ||
CR456588.1 | - | 5 | 575 |