Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013244.3 | NP_037376.2 | 7 | 949 | ||
XM_005268776.4 | XP_005268833.1 | 6 | 706 | |||
XM_005268779.3 | XP_005268836.1 | 5 | 1474 | |||
XM_011538150.2 | XP_011536452.1 | 6 | 773 | |||
XM_011538151.2 | XP_011536453.1 | 6 | 689 | |||
XM_011538156.2 | XP_011536458.1 | 9 | 1981 | |||
XM_011538157.2 | XP_011536459.1 | 8 | 2640 | |||
XM_017019143.1 | XP_016874632.1 | 6 | 1498 | |||
XM_017019144.1 | XP_016874633.1 | 6 | 679 | |||
XM_017019145.1 | XP_016874634.1 | 7 | 814 | |||
XM_017019146.1 | XP_016874635.1 | 7 | 811 | |||
XM_017019147.1 | XP_016874636.1 | 6 | 789 | |||
XM_017019148.1 | XP_016874637.1 | 8 | 1773 | |||
XM_017019149.1 | XP_016874638.1 | 7 | 1695 | |||
XM_017019150.1 | XP_016874639.1 | 8 | 1077 | |||
GenBank mRNA | AK127727.1 | - | 8 | 1688 | ||
AK299253.1 | - | 6 | 706 | |||
BC026068.1 | - | 6 | 713 | |||
BC064141.1 | - | 3 | 490 | 30378187 |