Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308053.1 | NP_001294982.1 | 10 | 896 | ||
NM_014353.4 | NP_055168.2 | 9 | 857 | |||
XM_011522448.1 | XP_011520750.1 | 9 | 915 | |||
XM_011522450.2 | XP_011520752.1 | 8 | 1674 | |||
GenBank mRNA | AB027137.1 | - | 8 | 528 | ||
AF498952.1 | - | 8 | 525 | |||
AK308560.1 | - | 9 | 1156 | |||
AK314110.1 | - | 10 | 896 | |||
AY646153.1 | - | 9 | 874 | |||
BC066913.1 | - | 9 | 863 | 5262795 |