Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195139.1 | NP_001182068.1 | 10 | 1284 | ||
NM_015386.2 | NP_056201.2 | 10 | 1284 | |||
XM_011522981.2 | XP_011521283.1 | 11 | 1444 | |||
GenBank mRNA | AB088369.1 | - | 11 | 1420 | ||
AK022874.1 | - | 10 | 1264 | |||
AK026435.1 | - | 6 | 578 | |||
AK096557.1 | - | 8 | 1002 | |||
AK293235.1 | - | 7 | 828 | |||
AK297557.1 | - | 11 | 1227 | |||
AL050101.1 | - | 6 | 586 | |||
BC006306.2 | - | 7 | 800 | |||
BC013347.2 | - | 6 | 592 | |||
BC027726.2 | - | 10 | 1258 | |||
BC072438.1 | - | 10 | 1264 | 6011802 | ||
BC128144.1 | - | 10 | 1261 | |||
BC128145.1 | - | 10 | 1261 | |||
BX648010.1 | - | 8 | 1952 |