Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014033.3 | NP_054752.3 | 2 | 825 | ||
XM_006719332.1 | XP_006719395.1 | 3 | 1179 | |||
GenBank mRNA | AF113007.1 | - | 2 | 499 | ||
AJ536233.1 | - | 2 | 547 | |||
AK023693.1 | - | 2 | 557 | |||
AK024409.1 | - | 2 | 590 | |||
AK225595.1 | - | 2 | 562 | |||
AK225689.1 | - | 2 | 561 | |||
AK293356.1 | - | 3 | 351 | |||
AK307071.1 | - | 3 | 417 | |||
AL050159.1 | - | 2 | 535 | |||
AY358797.1 | - | 2 | 593 | |||
BC004492.2 | - | 2 | 542 | 3689692 | ||
BC008180.2 | - | 2 | 543 | 2900478 |