Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145804.2 | NP_665803.2 | 4, 5 | 1832 | ||
GenBank mRNA | AK091560.1 | - | 1, 2 | 50 | ||
AK095632.1 | - | 4, 5 | 1079 | |||
AK291744.1 | - | 4, 5 | 1833 | |||
BC093968.1 | - | 4, 5 | 944 | 7939813 |