Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014034.2 | NP_054753.1 | 2 | 504 | ||
GenBank mRNA | AB028628.1 | - | 2 | 330 | ||
AF151856.1 | - | 4 | 781 | |||
AF161495.1 | - | 4 | 781 | |||
AF279306.1 | - | 2 | 316 | |||
AK025738.1 | - | 2 | 361 | |||
AL050261.1 | - | 2 | 359 | |||
BC010878.1 | - | 2 | 343 | 3546858 | ||
CR457347.1 | - | 2 | 167 | |||
DA902777.1 | - | 2 | 504 |