Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014034.2 | NP_054753.1 | 3 | 630 | ||
GenBank mRNA | AB028628.1 | - | 3 | 456 | ||
AF151856.1 | - | 5 | 907 | |||
AF161495.1 | - | 5 | 907 | |||
AF279306.1 | - | 3 | 442 | |||
AK025738.1 | - | 3 | 487 | |||
AL050261.1 | - | 3 | 485 | |||
BC010878.1 | - | 3 | 469 | 3546858 | ||
CR457347.1 | - | 3 | 293 |