Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015395.2 | NP_056210.1 | 17 | 2734 | ||
XM_005250253.3 | XP_005250310.1 | 17 | 2706 | |||
XM_017011937.1 | XP_016867426.1 | 16 | 2616 | |||
GenBank mRNA | AB037779.1 | - | 15 | 2310 | ||
AK027072.1 | - | 6 | 637 | |||
AK057048.1 | - | 16 | 2461 | |||
AK292996.1 | - | 17 | 2636 | |||
AL117495.1 | - | 6 | 1631 | |||
BC012529.2 | - | 5 | 557 | |||
BC053591.1 | - | 17 | 2639 | 5502429 |