Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2463
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
9
1383
NM_001198626.1
NP_001185555.1
7
1241
NM_001198627.1
NP_001185556.1
8
1301
NM_001198628.1
NP_001185557.1
6
898
NM_001198629.1
NP_001185558.1
6
1024
NM_001198630.1
NP_001185559.1
7
1084
NM_001198631.1
NP_001185560.1
6
1097
NM_001198632.1
NP_001185561.1
6
1015
NM_001198633.1
NP_001185562.1
5
986
NM_001198634.1
NP_001185563.1
5
924
NM_001198679.1
NP_001185608.1
6
1068
NM_004349.3
NP_004340.1
5
1034
NM_175634.2
NP_783552.1
6
1159
NM_175635.2
NP_783553.1
5
851
NM_175636.2
NP_783554.1
4
933
XM_006716676.3
XP_006716739.1
7
968
XM_011517351.2
XP_011515653.1
7
1013
XM_011517352.2
XP_011515654.1
7
1253
XM_011517353.2
XP_011515655.1
6
961
XM_017013930.1
XP_016869419.1
6
1269
XM_017013931.1
XP_016869420.1
5
791
XM_017013932.1
XP_016869421.1
7
1156
XM_017013933.1
XP_016869422.1
8
1220
XM_017013934.1
XP_016869423.1
8
1328
XM_017013935.1
XP_016869424.1
7
1133
XM_017013936.1
XP_016869425.1
6
1227
XM_017013937.1
XP_016869426.1
7
1053
XM_017013938.1
XP_016869427.1
6
917
GenBank mRNA
AK297616.1
-
5
748
AK312592.1
-
9
1145
BC005850.1
-
5
813
2961112
BC067078.1
-
5
804
2964656
BC139783.1
-
1
1626
BT009871.1
-
4
593
CR456792.1
-
4
623
D14289.1
-
4
890
D14821.1
-
5
788
D43638.1
-
5
1034
KF946505.1
-
5
652
KF946507.1
-
5
652
KF946508.1
-
5
652
KF946510.1
-
5
652
KF946511.1
-
5
638
KF946512.1
-
6
1942
KF946513.1
-
5
2322
KF946514.1
-
5
638
KF946515.1
-
4
878
KF946516.1
-
4
878
KF946517.1
-
4
878
KF946518.1
-
4
878
KF946519.1
-
1
300
KF946520.1
-
1
300
KF946521.1
-
1
300
KF946522.1
-
1
300
KF946523.1
-
1
300
X79990.1
-
4
616
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108657, XM_006237903, XM_008763549, XM_006237904, XM_008763548, and others.
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0