Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083885.2 | NP_001077354.2 | 11 | 1944 | ||
NM_001173425.1 | NP_001166896.1 | 11 | 3105 | |||
NM_015404.3 | NP_056219.3 | 11 | 3108 | |||
XM_011518484.1 | XP_011516786.1 | 12 | 3194 | |||
XM_011518485.1 | XP_011516787.1 | 13 | 3042 | |||
XM_011518486.1 | XP_011516788.1 | 12 | 3191 | |||
XM_011518487.1 | XP_011516789.1 | 11 | 3068 | |||
XM_011518488.1 | XP_011516790.1 | 10 | 2951 | |||
XM_011518495.1 | XP_011516797.1 | 9 | 1446 | |||
GenBank mRNA | AB040959.1 | - | 11 | 2622 | ||
AK022854.1 | - | 7 | 1822 | |||
AL110228.1 | - | 11 | 1902 | |||
BC011918.2 | - | 2 | 217 | |||
BC014524.1 | - | 3605 | ||||
BC136416.1 | - | 11 | 2854 | 9020404 | ||
BC142614.1 | - | 11 | 3108 | 40147287 | ||
BC142684.1 | - | 11 | 3083 | 40148465 |