Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083885.2 | NP_001077354.2 | 11, 12 | 2025 | ||
NM_001173425.1 | NP_001166896.1 | 11, 12 | 3186 | |||
NM_015404.3 | NP_056219.3 | 11, 12 | 3189 | |||
XM_011518484.1 | XP_011516786.1 | 12, 13 | 3275 | |||
XM_011518485.1 | XP_011516787.1 | 13, 14 | 3123 | |||
XM_011518486.1 | XP_011516788.1 | 12, 13 | 3272 | |||
XM_011518487.1 | XP_011516789.1 | 11, 12 | 3149 | |||
XM_011518488.1 | XP_011516790.1 | 10, 11 | 3032 | |||
XM_011518495.1 | XP_011516797.1 | 9, 10 | 1527 | |||
GenBank mRNA | AB040959.1 | - | 11, 12 | 2703 | ||
AK022854.1 | - | 7, 8 | 1903 | |||
AL110228.1 | - | 11, 12 | 1983 | |||
BC011918.2 | - | 2, 3 | 298 | |||
BC014524.1 | - | 3686 | ||||
BC136416.1 | - | 11, 12 | 2935 | 9020404 | ||
BC142614.1 | - | 11, 12 | 3189 | 40147287 | ||
BC142684.1 | - | 11, 12 | 3164 | 40148465 |