Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD14A-ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316331.1 | NP_001303260.1 | 2 | 348 | ||
GenBank mRNA | AK298086.1 | - | 2 | 324 |
Gene Symbol: ABHD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015407.4 | NP_056222.2 | 2 | 348 | ||
GenBank mRNA | AK308592.1 | - | 2 | 272 | ||
AL050015.1 | - | 2 | 260 | |||
AY358201.1 | - | 2 | 305 | |||
BC002571.2 | - | 2 | 306 | 3143135 | ||
CN256406.1 | - | 2 | 352 |