Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 10 | 1776 | ||
NM_001079881.1 | NP_001073350.1 | 10 | 1640 | |||
NM_001079882.1 | NP_001073351.1 | 8 | 1377 | |||
NM_016457.4 | NP_057541.2 | 9 | 2055 | |||
XM_005258716.2 | XP_005258773.2 | 8 | 1033 | |||
GenBank mRNA | AF309082.1 | - | 9 | 1337 | ||
AK074673.1 | - | 8 | 1016 | |||
AK075266.1 | - | 8 | 1126 | |||
AK092896.1 | - | 9 | 1427 | |||
AK095884.1 | - | 10 | 1319 | |||
AK300339.1 | - | 10 | 1590 | |||
AK303629.1 | - | 8 | 1031 | |||
AL050147.1 | - | 3 | 292 | |||
BC025307.1 | - | 5 | 626 |