Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LETMD1 SIRNA Assays ›
Gene Symbol:
LETMD1
Gene Name:
LETM1 domain containing 1
Gene Aliases:
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location:
Chr.12: 51048230 - 51068984 on Build GRCh38
UniGene ID:
Hs.655272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24658
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LETMD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243689.1
NP_001230618.1
6
779
NM_015416.4
NP_056231.3
6
740
NR_045017.1
6
615
NR_045018.1
5
624
NR_045019.1
4
541
XM_006719335.1
XP_006719398.1
5
562
XM_006719336.1
XP_006719399.1
5
450
XM_006719339.1
XP_006719402.1
6
678
XM_006719340.1
XP_006719403.1
5
499
XM_011538161.2
XP_011536463.1
6
747
XM_011538162.2
XP_011536464.1
6
748
XM_011538163.2
XP_011536465.1
6
749
XM_011538164.2
XP_011536466.1
5
597
XM_017019151.1
XP_016874640.1
6
749
XM_017019152.1
XP_016874641.1
6
715
XM_017019153.1
XP_016874642.1
5
599
XM_017019154.1
XP_016874643.1
5
562
XM_017019155.1
XP_016874644.1
6
587
XM_017019156.1
XP_016874645.1
4
447
XM_017019157.1
XP_016874646.1
4
409
XM_017019158.1
XP_016874647.1
5
450
XM_017019163.1
XP_016874652.1
4
409
XM_017019164.1
XP_016874653.1
5
616
XM_017019165.1
XP_016874654.1
6
678
XM_017019166.1
XP_016874655.1
5
499
GenBank mRNA
AF195651.2
-
6
712
AF315598.1
-
6
576
AF329277.1
-
5
623
AK000639.1
-
6
727
AK057866.1
-
6
685
AK127540.1
-
6
740
AK225836.1
-
6
698
AK301898.1
-
5
550
AK301955.1
-
6
615
AK315119.1
-
6
707
AY259835.1
-
4
483
BC064943.1
-
6
686
6009854
BX343550.2
-
4
482
DA682074.1
-
5
499
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Process(es)
protein binding
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-kq4fm:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline