Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LETMD1 SIRNA Assays ›
Gene Symbol:
LETMD1
Gene Name:
LETM1 domain containing 1
Gene Aliases:
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location:
Chr.12: 51048230 - 51068984 on Build GRCh38
UniGene ID:
Hs.655272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24660
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LETMD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243689.1
NP_001230618.1
7
941
NM_001300765.1
NP_001287694.1
4
530
NM_015416.4
NP_056231.3
7
902
NR_045017.1
7
777
NR_045018.1
6
786
NR_045019.1
5
703
NR_045020.1
3
414
XM_006719335.1
XP_006719398.1
6
724
XM_006719336.1
XP_006719399.1
6
612
XM_006719339.1
XP_006719402.1
7
840
XM_006719340.1
XP_006719403.1
6
661
XM_011538161.2
XP_011536463.1
7
909
XM_011538162.2
XP_011536464.1
7
910
XM_011538163.2
XP_011536465.1
7
911
XM_011538164.2
XP_011536466.1
6
759
XM_011538165.2
XP_011536467.1
4
537
XM_011538167.1
XP_011536469.1
2
331
XM_017019151.1
XP_016874640.1
7
911
XM_017019152.1
XP_016874641.1
7
877
XM_017019153.1
XP_016874642.1
6
761
XM_017019154.1
XP_016874643.1
6
724
XM_017019155.1
XP_016874644.1
7
749
XM_017019156.1
XP_016874645.1
5
609
XM_017019157.1
XP_016874646.1
5
571
XM_017019158.1
XP_016874647.1
6
612
XM_017019159.1
XP_016874648.1
4
540
XM_017019160.1
XP_016874649.1
4
502
XM_017019161.1
XP_016874650.1
4
508
XM_017019162.1
XP_016874651.1
4
478
XM_017019163.1
XP_016874652.1
5
571
XM_017019164.1
XP_016874653.1
6
778
XM_017019165.1
XP_016874654.1
7
840
XM_017019166.1
XP_016874655.1
6
661
XM_017019167.1
XP_016874656.1
4
502
XM_017019168.1
XP_016874657.1
4
502
GenBank mRNA
AF195651.2
-
7
874
AF315598.1
-
7
738
AF329277.1
-
6
785
AK000639.1
-
7
889
AK057866.1
-
7
847
AK127540.1
-
7
902
AK225836.1
-
7
860
AK301898.1
-
6
712
AK301955.1
-
7
777
AK304707.1
-
3
386
AK315119.1
-
7
869
AL050286.1
-
2
394
AY259835.1
-
5
645
AY259836.1
-
3
356
BC000395.2
-
2
151
BC019274.1
-
2
151
BC064943.1
-
7
848
6009854
BM549678.1
-
386
BX343550.2
-
5
644
BX376670.2
-
4
471
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Process(es)
protein binding
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0