Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199809.1 | NP_001186738.1 | 18 | 2586 | ||
NM_001199811.1 | NP_001186740.1 | 19 | 2691 | |||
NM_001199812.1 | NP_001186741.1 | 19 | 2673 | |||
NM_015434.3 | NP_056249.1 | 19 | 2733 | |||
NR_037667.1 | 20 | 2767 | ||||
XM_011509396.2 | XP_011507698.1 | 20 | 2726 | |||
GenBank mRNA | AK001363.1 | - | 19 | 2533 | ||
AK001598.1 | - | 19 | 2615 | |||
AK022509.1 | - | 19 | 2593 | |||
AK022589.1 | - | 11 | 1400 | |||
AK297225.1 | - | 18 | 2465 | |||
AL117576.1 | - | 8 | 1007 | |||
AL133049.2 | - | 14 | 1902 | |||
BC020523.2 | - | 19 | 2567 | 3917030 | ||
BC030716.1 | - | 19 | 2588 | 3909231 | ||
BC033918.1 | - | 19 | 2592 | 3920829 |