Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009922.2 | NP_001009922.1 | 8 | 791 | ||
NM_001278536.1 | NP_001265465.1 | 8 | 698 | |||
NM_001278537.1 | NP_001265466.1 | 7 | 671 | |||
NM_001278538.1 | NP_001265467.1 | 9 | 774 | |||
NM_001278539.1 | NP_001265468.1 | 9 | 856 | |||
NM_015436.3 | NP_056251.2 | 9 | 818 | |||
NR_037913.1 | 9 | 846 | ||||
NR_037914.1 | 9 | 842 | ||||
NR_103723.1 | 9 | 816 | ||||
NR_103724.1 | 9 | 780 | ||||
NR_103725.1 | 7 | 565 | ||||
XM_011531838.2 | XP_011530140.1 | 9 | 787 | |||
XM_011531839.2 | XP_011530141.1 | 9 | 975 | |||
GenBank mRNA | AB209072.1 | - | 9 | 681 | ||
AF247041.1 | - | 9 | 674 | |||
AF255666.1 | - | 9 | 700 | |||
AF305424.1 | - | 9 | 708 | |||
AK091501.1 | - | 9 | 789 | |||
AK308754.1 | - | 8 | 551 | |||
AK308848.1 | - | 6 | 600 | |||
AK309555.1 | - | 9 | 689 | |||
AL050144.1 | - | 8 | 575 | |||
AY888047.1 | - | 9 | 662 | |||
BC015464.1 | - | 4 | 243 | |||
BC047393.1 | - | 9 | 818 | 5301408 | ||
BG703058.1 | - | 9 | 687 | |||
BI913695.1 | - | 7 | 483 | |||
BP314571.1 | - | 7 | 538 | |||
BQ959587.1 | - | 9 | 647 | |||
CF597260.1 | - | 7 | 387 | |||
GQ250944.1 | - | 9 | 674 | |||
GQ250945.1 | - | 8 | 647 | |||
GU937000.1 | - | 9 | 702 | |||
JF691538.1 | - | 9 | 674 |