Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015448.1 | NP_056263.1 | 1, 2 | 78 | ||
GenBank mRNA | AK290154.1 | - | 1, 2 | 79 | ||
AY532267.1 | - | 1, 2 | 65 | |||
BC001082.2 | - | 1, 2 | 78 | 2907119 | ||
BC031695.1 | - | 1, 2 | 101 | 5166605 |