Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015448.1 | NP_056263.1 | 4, 5 | 408 | ||
GenBank mRNA | AL110240.1 | - | 3, 4 | 329 | ||
AY532267.1 | - | 4, 5 | 395 | |||
BC001082.2 | - | 4, 5 | 408 | 2907119 | ||
BC031695.1 | - | 4, 5 | 431 | 5166605 |