Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001756.3 | NP_001747.2 | 2 | 470 | ||
GenBank mRNA | AK225591.1 | - | 2 | 452 | ||
AK290821.1 | - | 2 | 455 | |||
AK313760.1 | - | 2 | 455 | |||
BC017462.1 | - | 2 | 424 | |||
BC056259.1 | - | 2 | 445 | 5810260 | ||
BC058021.1 | - | 2 | 454 | 6714173 | ||
J02943.1 | - | 1 | 401 |