Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284423.1 | NP_001271352.1 | 14 | 2442 | ||
NM_001284424.1 | NP_001271353.1 | 13 | 2436 | |||
NM_001284425.1 | NP_001271354.1 | 13 | 2494 | |||
NM_001284426.1 | NP_001271355.1 | 12 | 2252 | |||
NM_015460.3 | NP_056275.2 | 14 | 2631 | |||
NR_104316.1 | 14 | 2594 | ||||
XM_011533575.1 | XP_011531877.1 | 14 | 2603 | |||
GenBank mRNA | AB083783.1 | - | 13 | 2344 | ||
AF396687.1 | - | 14 | 2480 | |||
AK125334.1 | - | 12 | 2118 | |||
AK126013.1 | - | 13 | 2390 | |||
AK294714.1 | - | 12 | 2242 | |||
AK312412.1 | - | 14 | 2441 | |||
AK316068.1 | - | 12 | 2252 | |||
AL050090.1 | - | 2 | 250 | |||
BC092512.1 | - | 14 | 2630 | 3093499 | ||
BC109311.1 | - | 13 | 2322 | 40036096 | ||
BC109312.1 | - | 14 | 2422 | 40036097 |