Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252156.1 | NP_001239085.1 | 27 | 4398 | ||
NM_015465.4 | NP_056280.2 | 27 | 4401 | |||
GenBank mRNA | AK074066.2 | - | 27 | 4373 | ||
AK315101.1 | - | 27 | 4384 | |||
AL117665.1 | - | 22 | 3463 | |||
AY063750.1 | - | 27 | 4318 | |||
BC008776.2 | - | 15 | 2559 | |||
BC014147.2 | - | 15 | 2559 | |||
BC113614.1 | - | 27 | 4357 | 8322666 | ||
BC143724.1 | - | 27 | 4354 | 9052238 |