Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304482.1 | NP_001291411.1 | 17, 18 | 1937 | ||
NM_015466.3 | NP_056281.1 | 18, 19 | 2065 | |||
XM_005265031.2 | XP_005265088.2 | 18, 19 | 2016 | |||
GenBank mRNA | AB025194.1 | - | 18, 19 | 2013 | ||
AB040904.2 | - | 18, 19 | 2065 | |||
AF290614.1 | - | 18, 19 | 2011 | |||
AK055157.1 | - | 18, 19 | 2036 | |||
AK289502.1 | - | 18, 19 | 2014 | |||
AK295877.1 | - | 18, 19 | 1985 | |||
AK295888.1 | - | 17, 18 | 1928 | |||
AK299906.1 | - | 17, 18 | 1908 | |||
BC027711.2 | - | 7, 8 | 1004 | |||
BC089042.1 | - | 18, 19 | 1983 | 6579163 |