Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013943.2 | NP_039234.1 | 6 | 910 | ||
GenBank mRNA | AF097330.1 | - | 6 | 765 | ||
AF109196.1 | - | 6 | 703 | |||
AK001624.1 | - | 6 | 798 | |||
AK095959.1 | - | 6 | 731 | |||
AK301466.1 | - | 6 | 708 | |||
AK307142.1 | - | 4 | 475 | |||
AK315051.1 | - | 6 | 747 | |||
AL117424.1 | - | 6 | 822 | |||
BC012444.1 | - | 6 | 775 | 3861372 | ||
CR533501.1 | - | 6 | 625 |