Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304538.1 | NP_001291467.1 | 6 | 809 | ||
NM_015475.4 | NP_056290.3 | 7 | 994 | |||
GenBank mRNA | AK074867.1 | - | 7 | 930 | ||
AK096187.1 | - | 3 | 1729 | |||
AK300023.1 | - | 6 | 738 | |||
AK302233.1 | - | 3 | 417 | |||
AL049943.1 | - | 2 | 241 | |||
BC038581.1 | - | 4 | 527 | |||
BC060860.1 | - | 7 | 907 | 30334952 | ||
BC136768.1 | - | 7 | 898 | 9020758 | ||
BC136770.1 | - | 7 | 898 | 9020760 | ||
CR936766.1 | - | 7 | 932 |