Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304538.1 | NP_001291467.1 | 7 | 1293 | ||
NM_015475.4 | NP_056290.3 | 8 | 1478 | |||
GenBank mRNA | AK027432.1 | - | 1 | 420 | ||
AK074867.1 | - | 8 | 1414 | |||
AK096187.1 | - | 4 | 2213 | |||
AK300023.1 | - | 7 | 1222 | |||
AK302233.1 | - | 4 | 901 | |||
AL049943.1 | - | 3 | 725 | |||
BC038581.1 | - | 5 | 1011 | |||
BC136768.1 | - | 8 | 1382 | 9020758 | ||
BC136770.1 | - | 8 | 1382 | 9020760 | ||
CR936766.1 | - | 8 | 1416 |