Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243286.1 | NP_001230215.1 | 4 | 1099 | ||
NM_001243288.1 | NP_001230217.1 | 4 | 1023 | |||
NM_015480.2 | NP_056295.1 | 4 | 1099 | |||
XM_011512663.1 | XP_011510965.1 | 5 | 1163 | |||
XM_017006123.1 | XP_016861612.1 | 5 | 1163 | |||
XM_017006124.1 | XP_016861613.1 | 5 | 820 | |||
XM_017006125.1 | XP_016861614.1 | 4 | 1023 | |||
XM_017006126.1 | XP_016861615.1 | 4 | 1094 | |||
XM_017006127.1 | XP_016861616.1 | 4 | 1186 | |||
GenBank mRNA | AF282874.1 | - | 4 | 840 | ||
AL050071.1 | - | 3 | 416 | |||
BC001336.1 | - | 2 | 107 | |||
BC067808.1 | - | 4 | 982 | 5285526 |