Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015483.2 | NP_056298.2 | 3, 4 | 993 | ||
XM_005249696.2 | XP_005249753.1 | 3, 4 | 849 | |||
XM_005249697.2 | XP_005249754.1 | 3, 4 | 582 | |||
XM_011515256.2 | XP_011513558.1 | 3, 4 | 728 | |||
XM_011515257.2 | XP_011513559.1 | 3, 4 | 462 | |||
GenBank mRNA | AF151831.1 | - | 2, 3 | 402 | ||
AK292802.1 | - | 3, 4 | 715 | |||
BC037887.2 | - | 3, 4 | 897 | 5274706 | ||
BC047107.1 | - | 3, 4 | 730 | 5275217 |