Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283014.1 | NP_001269943.1 | 4 | 465 | ||
NM_001283015.1 | NP_001269944.1 | 4 | 465 | |||
NM_001283016.1 | NP_001269945.1 | 5 | 537 | |||
NM_001283017.1 | NP_001269946.1 | 5 | 537 | |||
NM_001283019.1 | NP_001269948.1 | 4 | 465 | |||
NM_015901.5 | NP_056985.3 | 4 | 465 | |||
NR_104264.1 | 4 | 465 | ||||
XM_017016033.1 | XP_016871522.1 | 4 | 385 | |||
GenBank mRNA | AK300980.1 | - | 5 | 390 | ||
AK303235.1 | - | 4 | 341 | |||
AK309755.1 | - | 4 | 318 | |||
AK316538.1 | - | 5 | 390 | |||
AL050114.1 | - | 1 | 25 | |||
AL521667.3 | - | 4 | 311 | |||
BC031224.1 | - | 4 | 385 | 4824935 | ||
BC046173.2 | - | 4 | 318 | 3873776 |