Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184717.1 | NP_001171646.1 | 4 | 1732 | ||
NM_001184718.1 | NP_001171647.1 | 4 | 1297 | |||
NM_015508.4 | NP_056323.2 | 4 | 1647 | |||
GenBank mRNA | AK097785.1 | - | 4 | 1476 | ||
AK291261.1 | - | 4 | 1482 | |||
AL080156.1 | - | 3 | 333 | |||
BC034397.1 | - | 4 | 1297 | |||
BX537965.1 | - | 4 | 1474 | |||
CR749647.1 | - | 4 | 1733 |