Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015509.3 | NP_056324.2 | 6, 7 | 760 | ||
NR_024260.1 | 5, 6 | 678 | ||||
GenBank mRNA | AK074858.1 | - | 6, 7 | 717 | ||
AK074880.1 | - | 6, 7 | 730 | |||
AK074923.1 | - | 6, 7 | 703 | |||
AK075013.1 | - | 6, 7 | 737 | |||
AK094805.1 | - | 1, 2 | 1302 | |||
AK290537.1 | - | 5, 6 | 603 | |||
AK314424.1 | - | 5, 6 | 612 | |||
AL050272.1 | - | 5, 6 | 604 | |||
BC002888.2 | - | 6, 7 | 662 | |||
BC067367.1 | - | 4, 5 | 432 | |||
BC084551.1 | - | 2, 3 | 271 | |||
BC110876.1 | - | 6, 7 | 694 | 5553942 |