Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015509.3 | NP_056324.2 | 3 | 355 | ||
NR_024260.1 | 3 | 355 | ||||
GenBank mRNA | AK074858.1 | - | 3 | 312 | ||
AK074880.1 | - | 3 | 325 | |||
AK074923.1 | - | 3 | 298 | |||
AK075013.1 | - | 3 | 332 | |||
AK290537.1 | - | 3 | 280 | |||
AK303675.1 | - | 3 | 298 | |||
AK314424.1 | - | 3 | 289 | |||
AL050272.1 | - | 3 | 284 | |||
BC002888.2 | - | 3 | 257 | |||
BC067367.1 | - | 1 | 27 | |||
BC110876.1 | - | 3 | 289 | 5553942 | ||
DA496321.1 | - | 3 | 355 |