Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015509.3 | NP_056324.2 | 4, 5 | 458 | ||
GenBank mRNA | AK074858.1 | - | 4, 5 | 415 | ||
AK074880.1 | - | 4, 5 | 428 | |||
AK074923.1 | - | 4, 5 | 401 | |||
AK075013.1 | - | 4, 5 | 435 | |||
BC002888.2 | - | 4, 5 | 360 | |||
BC067367.1 | - | 2, 3 | 130 | |||
BC110876.1 | - | 4, 5 | 392 | 5553942 | ||
DA496321.1 | - | 4, 5 | 458 |