Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282433.1 | NP_001269362.1 | 4 | 851 | ||
NM_015515.4 | NP_056330.3 | 5 | 1379 | |||
XM_005257200.4 | XP_005257257.1 | 4 | 432 | |||
XM_011524595.1 | XP_011522897.1 | 4 | 422 | |||
GenBank mRNA | AF102848.1 | - | 4 | 807 | ||
AK002047.1 | - | 4 | 791 | |||
AK225866.1 | - | 4 | 1162 | |||
AK289449.1 | - | 4 | 874 | |||
AK302109.1 | - | 4 | 660 | |||
AK310880.1 | - | 4 | 963 | |||
AK314604.1 | - | 4 | 591 | |||
AL117538.1 | - | 3 | 635 | |||
BC028356.2 | - | 5 | 1159 | 4838347 | ||
CV373638.1 | - | 1 | 67 |