Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSKU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258210.1 | NP_001245139.1 | 2 | 534 | ||
NM_001318477.1 | NP_001305406.1 | 2 | 1431 | |||
NM_001318478.1 | NP_001305407.1 | 2 | 631 | |||
NM_001318479.1 | NP_001305408.1 | 2 | 442 | |||
NM_015516.3 | NP_056331.2 | 2 | 577 | |||
GenBank mRNA | AF191019.1 | - | 1 | 418 | ||
AK075476.1 | - | 2 | 489 | |||
AK091475.1 | - | 3 | 394 | |||
AK126536.1 | - | 2 | 1431 | |||
AK313405.1 | - | 2 | 490 | |||
AL110276.1 | - | 1 | 73 | |||
AY358317.1 | - | 2 | 468 | |||
BC020975.1 | - | 2 | 455 | 3848274 | ||
BT007440.1 | - | 1 | 405 | |||
DA879213.1 | - | 2 | 442 |