Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099436.3 | NP_001092906.3 | 9 | 1087 | ||
NM_001284364.2 | NP_001271293.2 | 9 | 1087 | |||
NM_001284365.2 | NP_001271294.1 | 9 | 1010 | |||
NR_104307.2 | 8 | 1268 | ||||
NR_104308.2 | 8 | 1268 | ||||
NR_104309.2 | 8 | 1268 | ||||
XM_005254289.2 | XP_005254346.1 | 10 | 1102 | |||
XM_017022068.1 | XP_016877557.1 | 10 | 1136 | |||
XM_017022069.1 | XP_016877558.1 | 10 | 1140 | |||
XM_017022070.1 | XP_016877559.1 | 10 | 1153 | |||
XM_017022071.1 | XP_016877560.1 | 10 | 1140 | |||
XM_017022072.1 | XP_016877561.1 | 9 | 1316 | |||
GenBank mRNA | AK075047.1 | - | 8 | 1177 | ||
AK293181.1 | - | 9 | 940 | |||
AK294241.1 | - | 9 | 1021 | |||
AK294245.1 | - | 9 | 1017 | |||
AK294251.1 | - | 9 | 1021 | |||
AK298879.1 | - | 9 | 944 | |||
AK299380.1 | - | 9 | 1014 | |||
AL117482.1 | - | 8 | 1403 | |||
AL360256.1 | - | 4 | 680 | |||
BC036117.1 | - | 8 | 1239 | |||
BC048289.1 | - | 8 | 1196 | |||
BC056423.1 | - | 8 | 1186 | |||
BC157884.1 | - | 9 | 1069 | 8862706 |