Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015523.3 | NP_056338.2 | 7 | 773 | ||
GenBank mRNA | AF151872.1 | - | 7 | 724 | ||
AK023498.1 | - | 3 | 1070 | |||
AK223200.1 | - | 7 | 688 | |||
AK312042.1 | - | 7 | 702 | |||
AL110239.1 | - | 7 | 707 | |||
BC003502.1 | - | 3 | 322 | |||
BC105024.1 | - | 7 | 694 | 8144027 | ||
BC105026.1 | - | 7 | 694 | 8144029 | ||
BC107887.1 | - | 683 | 6663083 | |||
BC143701.1 | - | 7 | 697 | 9052215 | ||
CR533500.1 | - | 7 | 630 |