Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015523.3 | NP_056338.2 | 2, 3 | 384 | ||
GenBank mRNA | AF151872.1 | - | 2, 3 | 335 | ||
AK223200.1 | - | 2, 3 | 299 | |||
AK312042.1 | - | 2, 3 | 313 | |||
AL110239.1 | - | 2, 3 | 318 | |||
BC105024.1 | - | 2, 3 | 305 | 8144027 | ||
BC105026.1 | - | 2, 3 | 305 | 8144029 | ||
BC107887.1 | - | 294 | 6663083 | |||
BC143701.1 | - | 2, 3 | 308 | 9052215 | ||
CR533500.1 | - | 2, 3 | 241 | |||
DB471224.1 | - | 2, 3 | 385 |