Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 10 | 1754 | ||
NM_015526.2 | NP_056341.1 | 11 | 1670 | |||
GenBank mRNA | AJ427922.1 | - | 11 | 1648 | ||
AK056445.1 | - | 9 | 1153 | |||
AK090732.1 | - | 11 | 1673 | |||
AK092354.1 | - | 7 | 917 | |||
AK094738.1 | - | 11 | 1668 | |||
AK289509.1 | - | 10 | 1722 | |||
AK294748.1 | - | 11 | 1541 | |||
AL117468.1 | - | 3 | 282 | |||
BC013116.1 | - | 11 | 1582 | 4214441 | ||
BC014486.1 | - | 8 | 1113 |