Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 3 | 675 | ||
NM_015526.2 | NP_056341.1 | 4 | 591 | |||
GenBank mRNA | AJ427922.1 | - | 4 | 569 | ||
AK056445.1 | - | 2 | 75 | |||
AK090732.1 | - | 4 | 594 | |||
AK094738.1 | - | 4 | 589 | |||
AK289509.1 | - | 3 | 643 | |||
AK294748.1 | - | 4 | 462 | |||
BC013116.1 | - | 4 | 503 | 4214441 | ||
BC014486.1 | - | 1 | 34 |