Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320356.1 | NP_001307285.1 | 6, 7 | 874 | ||
NM_001320357.1 | NP_001307286.1 | 5, 6 | 764 | |||
NM_001320358.1 | NP_001307287.1 | 5, 6 | 634 | |||
NM_001320359.1 | NP_001307288.1 | 6, 7 | 790 | |||
NM_001320360.1 | NP_001307289.1 | 5, 6 | 1020 | |||
NM_001320361.1 | NP_001307290.1 | 6, 7 | 1004 | |||
NM_001320362.1 | NP_001307291.1 | 5, 6 | 584 | |||
NM_001320363.1 | NP_001307292.1 | 5, 6 | 714 | |||
NM_001320364.1 | NP_001307293.1 | 4, 5 | 643 | |||
NM_001320365.1 | NP_001307294.1 | 6, 7 | 954 | |||
NM_015528.2 | NP_056343.1 | 5, 6 | 1150 | |||
XM_017024427.1 | XP_016879916.1 | 4, 5 | 890 | |||
GenBank mRNA | AK025329.1 | - | 5, 6 | 823 | ||
AL050060.1 | - | 3, 4 | 300 | |||
AL537131.3 | - | 6, 7 | 678 | |||
AL834284.1 | - | 4, 5 | 672 | |||
AY203930.1 | - | 4, 5 | 650 | |||
AY960626.1 | - | 4, 5 | 317 | |||
BC010139.2 | - | 4, 5 | 548 | 4128232 | ||
BC013222.1 | - | 4, 5 | 604 | |||
BI825561.1 | - | 5, 6 | 486 | |||
BI830686.1 | - | 5, 6 | 664 | |||
CB150714.1 | - | 5, 6 | 536 | |||
CR457340.1 | - | 4, 5 | 374 |