Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015530.4 | NP_056345.3 | 1, 2 | 877 | ||
GenBank mRNA | AK023082.1 | - | 1, 2 | 149 | ||
AK027349.1 | - | 1, 2 | 76 | |||
AK125761.1 | - | 1, 2 | 877 | |||
AK293640.1 | - | 1, 2 | 150 | |||
AK298020.1 | - | 1, 2 | 131 | |||
AK315508.1 | - | 1, 2 | 178 | |||
BC007770.2 | - | 1, 2 | 111 | 4298251 | ||
BG034906.1 | - | 1, 2 | 202 |